Assay Design



Gene Transcripts

Gene Symbol
NRIP2
Entrez Gene ID
101118863
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 227738648 - 227746632 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006958.4 XP_004007007.4 3 - 4 607 64

View Full Product Details