Assay Design



Gene Transcripts

Gene Symbol
GAST
Entrez Gene ID
101119230
Gene Name
gastrin
Gene Aliases
GAS
Chromosome Location
Chr.11: 21207670 - 21208306 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975447.1 XP_027831248.1 1 - 2 221 110

View Full Product Details