Assay Design



Gene Transcripts

Gene Symbol
RPL11
Entrez Gene ID
101120638
Gene Name
ribosomal protein L11
Gene Aliases
-
Chromosome Location
Chr.2: 258726294 - 258730104 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093687.2 XP_014949173.1 3 - 4 319 96

View Full Product Details