Assay Design



Gene Transcripts

Gene Symbol
ACTL7A
Entrez Gene ID
101115937
Gene Name
actin like 7A
Gene Aliases
-
Chromosome Location
Chr.2: 14854549 - 14855865 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092919.2 XP_014948405.1 1 - 1 1061 65

View Full Product Details