Assay Design



Gene Transcripts

Gene Symbol
NANOG
Entrez Gene ID
100302345
Gene Name
Nanog homeobox
Gene Aliases
-
Chromosome Location
Chr.3: 222316615 - 222321231 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006901.4 XP_004006950.3 2 - 3 400 105
XM_015094959.2 XP_014950445.1 2 - 3 400 105
GenBank mRNA FJ970651.1 - 1 - 2 255 105

View Full Product Details