Assay Design



Gene Transcripts

Gene Symbol
KLHL5
Entrez Gene ID
101112731
Gene Name
kelch like family member 5
Gene Aliases
-
Chromosome Location
Chr.6: 63803601 - 63902455 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009770.4 XP_004009819.1 1 - 2 3324 116

View Full Product Details