Assay Design



Gene Transcripts

Gene Symbol
TUBAL3
Entrez Gene ID
101107301
Gene Name
tubulin alpha like 3
Gene Aliases
-
Chromosome Location
Chr.13: 44696378 - 44705793 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012188597.3 XP_012043987.1 2 - 3 304 91

View Full Product Details