Assay Design



Gene Transcripts

Gene Symbol
LOC101123662
Entrez Gene ID
101123662
Gene Name
neuronal acetylcholine receptor subunit alpha-10
Gene Aliases
CHRNA10
Chromosome Location
Chr.15: 54422536 - 54427836 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100802.2 XP_014956288.2 4 - 5 1018 106

Gene Symbol
LOC114118428
Entrez Gene ID
114118428
Gene Name
neuronal acetylcholine receptor subunit alpha-10
Gene Aliases
-
Chromosome Location
Chr.15: 54458211 - 54463746 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979455.1 XP_027835256.1 4 - 5 1766 106

Gene Symbol
LOC114118504
Entrez Gene ID
114118504
Gene Name
neuronal acetylcholine receptor subunit alpha-10-like
Gene Aliases
-
Chromosome Location
Chr.15: 54433228 - 54437243 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979872.1 XP_027835673.1 4 - 5 945 106

View Full Product Details