Assay Design



Gene Transcripts

Gene Symbol
HES5
Entrez Gene ID
101103410
Gene Name
hes family bHLH transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.12: 54098863 - 54099701 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013770.4 XP_004013819.1 1 - 2 88 60

View Full Product Details