Assay Design



Gene Transcripts

Gene Symbol
PDF
Entrez Gene ID
101112923
Gene Name
peptide deformylase, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.14: 39246025 - 39248073 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015605.4 XP_004015654.2 1 - 2 728 62

View Full Product Details