Assay Design



Gene Transcripts

Gene Symbol
HAO2
Entrez Gene ID
101110803
Gene Name
hydroxyacid oxidase 2
Gene Aliases
-
Chromosome Location
Chr.1: 101895564 - 101925651 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002387.4 XP_004002436.2 2 - 3 429 87
XM_012179575.3 XP_012034965.2 2 - 3 406 87
XM_012179579.3 XP_012034969.2 3 - 4 512 87

View Full Product Details