Assay Design



Gene Transcripts

Gene Symbol
CLDN11
Entrez Gene ID
101113649
Gene Name
claudin 11
Gene Aliases
-
Chromosome Location
Chr.1: 236124550 - 236139391 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003841.4 XP_004003890.4 2 - 3 592 90

View Full Product Details