Assay Design



Gene Transcripts

Gene Symbol
GSG1
Entrez Gene ID
101109690
Gene Name
germ cell associated 1
Gene Aliases
-
Chromosome Location
Chr.3: 217051839 - 217058431 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965844.1 XP_027821645.1 2 - 3 492 86

View Full Product Details