Assay Design



Gene Transcripts

Gene Symbol
SH2D4B
Entrez Gene ID
101109766
Gene Name
SH2 domain containing 4B
Gene Aliases
-
Chromosome Location
Chr.25: 37024122 - 37101490 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962301.1 XP_027818102.1 4 - 5 656 65

View Full Product Details