Assay Design



Gene Transcripts

Gene Symbol
ANKK1
Entrez Gene ID
101109481
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 26473151 - 26487012 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979116.1 XP_027834917.1 7 - 8 905 75

View Full Product Details