Assay Design



Gene Transcripts

Gene Symbol
KRT2
Entrez Gene ID
101109688
Gene Name
keratin 2
Gene Aliases
-
Chromosome Location
Chr.3: 143271718 - 143279600 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006321.4 XP_004006370.1 6 - 7 1243 62

View Full Product Details