Assay Design



Gene Transcripts

Gene Symbol
MMP19
Entrez Gene ID
101113485
Gene Name
matrix metallopeptidase 19
Gene Aliases
-
Chromosome Location
Chr.3: 175328456 - 175334324 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006603.4 XP_004006652.1 1 - 2 222 84
XM_012174781.3 XP_012030171.1 1 - 2 224 84

View Full Product Details