Assay Design



Gene Transcripts

Gene Symbol
CASS4
Entrez Gene ID
101105178
Gene Name
Cas scaffold protein family member 4
Gene Aliases
-
Chromosome Location
Chr.13: 61821627 - 61857170 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977022.1 XP_027832823.1 5 - 6 2778 99

View Full Product Details