Assay Design



Gene Transcripts

Gene Symbol
CDSN
Entrez Gene ID
101109128
Gene Name
corneodesmosin
Gene Aliases
-
Chromosome Location
Chr.20: 29766480 - 29771111 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958867.1 XP_027814668.1 1 - 2 95 95

View Full Product Details