Assay Design



Gene Transcripts

Gene Symbol
PROX2
Entrez Gene ID
101108394
Gene Name
prospero homeobox 2
Gene Aliases
-
Chromosome Location
Chr.7: 89862407 - 89871331 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015097183.2 XP_014952669.2 3 - 4 1596 114
XM_027972028.1 XP_027827829.1 2 - 3 915 114

View Full Product Details