Assay Design



Gene Transcripts

Gene Symbol
C1S
Entrez Gene ID
101107688
Gene Name
complement C1s
Gene Aliases
-
Chromosome Location
Chr.3: 224176808 - 224188167 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006917.4 XP_004006966.1 11 - 12 1592 107
XM_012175494.3 XP_012030884.1 11 - 12 1621 107

View Full Product Details