Assay Design



Gene Transcripts

Gene Symbol
GFRA4
Entrez Gene ID
101111478
Gene Name
GDNF family receptor alpha 4
Gene Aliases
-
Chromosome Location
Chr.13: 53584675 - 53589623 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099464.2 XP_014954950.2 1 - 2 224 60

View Full Product Details