Assay Design



Gene Transcripts

Gene Symbol
RBP5
Entrez Gene ID
101107181
Gene Name
retinol binding protein 5
Gene Aliases
-
Chromosome Location
Chr.3: 224073675 - 224079081 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012175482.2 XP_012030872.1 4 - 5 715 62
XM_027967920.1 XP_027823721.1 3 - 4 499 62

View Full Product Details