Assay Design



Gene Transcripts

Gene Symbol
DDX19A
Entrez Gene ID
101115151
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19B
Chromosome Location
Chr.14: 1260712 - 1280469 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100363.2 XP_014955849.2 6 - 7 589 60

View Full Product Details