Assay Design



Gene Transcripts

Gene Symbol
KCNK1
Entrez Gene ID
101115679
Gene Name
potassium two pore domain channel subfamily K member 1
Gene Aliases
-
Chromosome Location
Chr.25: 6132003 - 6202842 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104331.2 XP_014959817.2 1 - 2 629 109

View Full Product Details