Assay Design



Gene Transcripts

Gene Symbol
FAM71E2
Entrez Gene ID
101120774
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
-
Chromosome Location
Chr.14: 66808143 - 66811961 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977985.1 XP_027833786.1 6 - 7 752 113

View Full Product Details