Assay Design



Gene Transcripts

Gene Symbol
OTOP3
Entrez Gene ID
101101904
Gene Name
otopetrin 3
Gene Aliases
-
Chromosome Location
Chr.11: 6797743 - 6804371 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975423.1 XP_027831224.1 4 - 5 920 57

View Full Product Details