Assay Design



Gene Transcripts

Gene Symbol
RBP1
Entrez Gene ID
101108977
Gene Name
retinol binding protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 272200576 - 272227074 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959868.1 XP_027815669.1 3 - 4 529 101

View Full Product Details