Assay Design



Gene Transcripts

Gene Symbol
AHRR
Entrez Gene ID
101109301
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
-
Chromosome Location
Chr.16: 78167473 - 78240974 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979965.1 XP_027835766.1 7 - 8 920 50

View Full Product Details