Assay Design



Gene Transcripts

Gene Symbol
SRARP
Entrez Gene ID
101111430
Gene Name
steroid receptor associated and regulated protein
Gene Aliases
C2H1orf64
Chromosome Location
Chr.2: 265583264 - 265588032 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012154155.3 XP_012009545.1 1 - 2 139 142

View Full Product Details