Assay Design



Gene Transcripts

Gene Symbol
FCRL4
Entrez Gene ID
101112622
Gene Name
Fc receptor like 4
Gene Aliases
-
Chromosome Location
Chr.1: 114221770 - 114246066 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977830.1 XP_027833631.1 2 - 3 353 75

View Full Product Details