Assay Design



Gene Transcripts

Gene Symbol
MS4A1
Entrez Gene ID
101113271
Gene Name
membrane spanning 4-domains A1
Gene Aliases
-
Chromosome Location
Chr.15: 89578583 - 89593658 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016513.4 XP_004016562.2 6 - 7 795 87

View Full Product Details