Assay Design



Gene Transcripts

Gene Symbol
BET1
Entrez Gene ID
101114348
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
-
Chromosome Location
Chr.4: 13154907 - 13169147 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007725.3 XP_004007774.1 2 - 3 302 112

View Full Product Details