Assay Design



Gene Transcripts

Gene Symbol
SLC46A1
Entrez Gene ID
101119738
Gene Name
solute carrier family 46 member 1
Gene Aliases
-
Chromosome Location
Chr.11: 43482840 - 43491142 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975238.1 XP_027831039.1 3 - 4 1423 99

View Full Product Details