Assay Design



Gene Transcripts

Gene Symbol
TVP23A
Entrez Gene ID
101108621
Gene Name
trans-golgi network vesicle protein 23 homolog A
Gene Aliases
FAM18A
Chromosome Location
Chr.24: 9711537 - 9743721 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020770.4 XP_004020819.1 2 - 3 695 72

View Full Product Details