Assay Design



Gene Transcripts

Gene Symbol
TNIP3
Entrez Gene ID
101114957
Gene Name
TNFAIP3 interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 5192955 - 5251787 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971373.1 XP_027827174.1 4 - 5 340 90

View Full Product Details