Assay Design



Gene Transcripts

Gene Symbol
C2H2orf80
Entrez Gene ID
101120472
Gene Name
chromosome 2 C2orf80 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 224358800 - 224395856 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964850.1 XP_027820651.1 7 - 8 633 115

View Full Product Details