Assay Design



Gene Transcripts

Gene Symbol
SLC16A13
Entrez Gene ID
101114882
Gene Name
solute carrier family 16 member 13
Gene Aliases
-
Chromosome Location
Chr.11: 36533343 - 36537161 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012625.4 XP_004012674.3 3 - 4 1503 100
XM_027975012.1 XP_027830813.1 3 - 4 970 100

View Full Product Details