Assay Design



Gene Transcripts

Gene Symbol
CLPS
Entrez Gene ID
101119081
Gene Name
colipase
Gene Aliases
-
Chromosome Location
Chr.20: 11450270 - 11452595 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018774.4 XP_004018823.1 1 - 2 120 89

View Full Product Details