Assay Design



Gene Transcripts

Gene Symbol
SUFU
Entrez Gene ID
101114993
Gene Name
SUFU negative regulator of hedgehog signaling
Gene Aliases
-
Chromosome Location
Chr.22: 25733050 - 25843036 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020324.4 XP_004020373.4 5 - 6 833 77
XM_015103389.2 XP_014958875.2 5 - 6 834 77
XM_015103390.2 XP_014958876.2 5 - 6 834 77

View Full Product Details