Assay Design



Gene Transcripts

Gene Symbol
PPY
Entrez Gene ID
443280
Gene Name
pancreatic polypeptide
Gene Aliases
-
Chromosome Location
Chr.11: 19244515 - 19245206 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975444.1 XP_027831245.1 1 - 2 191 63
GenBank mRNA AY427976.1 - 1 - 2 163 63

View Full Product Details