Assay Design



Gene Transcripts

Gene Symbol
FAM53B
Entrez Gene ID
101104621
Gene Name
family with sequence similarity 53 member B
Gene Aliases
-
Chromosome Location
Chr.22: 47623707 - 47751175 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103490.2 XP_014958976.2 4 - 5 1420 81
XM_027960195.1 XP_027815996.1 4 - 5 1164 81
XM_027960196.1 XP_027815997.1 3 - 4 2317 81

View Full Product Details