Assay Design



Gene Transcripts

Gene Symbol
NENF
Entrez Gene ID
101103662
Gene Name
neudesin neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.17: 38482369 - 38489650 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956303.1 XP_027812104.1 3 - 4 395 69

View Full Product Details