Assay Design



Gene Transcripts

Gene Symbol
YJU2
Entrez Gene ID
101112814
Gene Name
YJU2 splicing factor homolog
Gene Aliases
CCDC94
Chromosome Location
Chr.5: 18276682 - 18291974 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012178071.3 XP_012033461.2 6 - 7 1496 57

View Full Product Details