Assay Design



Gene Transcripts

Gene Symbol
R3HDML
Entrez Gene ID
101106099
Gene Name
R3H domain containing like
Gene Aliases
-
Chromosome Location
Chr.13: 75893378 - 75902881 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099892.2 XP_014955378.2 1 - 2 250 99

View Full Product Details