Assay Design



Gene Transcripts

Gene Symbol
CES3
Entrez Gene ID
101105682
Gene Name
carboxylesterase 3
Gene Aliases
-
Chromosome Location
Chr.14: 37434534 - 37447384 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012189895.3 XP_012045285.2 10 - 11 1353 77
XM_015100471.2 XP_014955957.2 10 - 11 1318 77

View Full Product Details