Assay Design



Gene Transcripts

Gene Symbol
ZNF7
Entrez Gene ID
101106019
Gene Name
zinc finger protein 7
Gene Aliases
-
Chromosome Location
Chr.9: 14549135 - 14558692 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012183412.3 XP_012038802.2 4 - 5 358 82
XM_012183415.3 XP_012038805.2 5 - 6 395 82
XM_027973346.1 XP_027829147.1 5 - 6 392 82

View Full Product Details