Assay Design



Gene Transcripts

Gene Symbol
MAGOH
Entrez Gene ID
101118828
Gene Name
mago homolog, exon junction complex subunit
Gene Aliases
-
Chromosome Location
Chr.1: 29369202 - 29378089 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004001993.4 XP_004002042.1 4 - 5 437 113

Gene Symbol
LOC114112996
Entrez Gene ID
114112996
Gene Name
protein mago nashi homolog
Gene Aliases
-
Chromosome Location
Chr.2: 89478410 - 89479085 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964560.1 XP_027820361.1 1 - 1 423 113

View Full Product Details