Assay Design



Gene Transcripts

Gene Symbol
MED30
Entrez Gene ID
101120338
Gene Name
mediator complex subunit 30
Gene Aliases
-
Chromosome Location
Chr.9: 66035963 - 66058923 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011987.3 XP_004012036.3 3 - 4 609 79

View Full Product Details