Assay Design



Gene Transcripts

Gene Symbol
CCDC38
Entrez Gene ID
101118778
Gene Name
coiled-coil domain containing 38
Gene Aliases
-
Chromosome Location
Chr.3: 177924632 - 177953013 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965962.1 XP_027821763.1 6 - 7 761 85

View Full Product Details